Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(p-Toluenesulfonyl)-1,2,4-triazole 98.0+%, TCI America™
SDP

Catalog No. T31871G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T31871G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T31871G Supplier TCI America Supplier No. T31871G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 13578-51-3
Melting Point 107°C
Color White
Molecular Formula C9H9N3O2S
Quantity 1 g
MDL Number MFCD00458591
Synonym 1-Tosyl-1,2,4-triazole
InChI Key GFWABQNSSIQCLB-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)S(=O)(=O)N1C=NC=N1
IUPAC Name 1-(4-methylbenzenesulfonyl)-1H-1,2,4-triazole
Molecular Weight (g/mol) 223.25
PubChem CID 3505424
Formula Weight 223.25
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(p-Toluenesulfonyl)-1,2,4-triazole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.