Learn More
Medchemexpress LLC 1-palmitoyl-2-arachidonoyl-sn-glycero-3-phosphocholine | 35418-58-7 | MFCD00171671 | 99.0% | C44H80NO8P | 10MG
SDP
Catalog No. 5000261431
Encompass_Preferred
1-palmitoyl-2-arachidonoyl-sn-glycero-3-phosphocholine (PAPC) is a phosphatidylcholine-class phospholipid containing a palmitoyl (16:0) chain at sn-1 and an arachidonoyl (20:4) chain at sn-2. It is provided as a high-purity research reagent and reference standard for studies in lipid biology, membrane structure, and signaling pathways.
  • High purity (99.0%) for analytical and research use.
  • Suitable for membrane biophysics and lipid signaling studies.
  • Typically supplied in small-mass vials or as a solution in chloroform for handling.
  • Well characterized: CAS 35418-58-7; formula C44H80NO8P.
Catalog No. 50-002-61431 Supplier Medchemexpress LLC Supplier No. HY14163610MG
Add to Cart
Add to Cart
1-palmitoyl-2-arachidonoyl-sn-glycero-3-phosphocholine (PAPC) is a phosphatidylcholine-class phospholipid containing a palmitoyl (16:0) chain at sn-1 and an arachidonoyl (20:4) chain at sn-2. It is provided as a high-purity research reagent and reference standard for studies in lipid biology, membrane structure, and signaling pathways.
  • High purity (99.0%) for analytical and research use.
  • Suitable for membrane biophysics and lipid signaling studies.
  • Typically supplied in small-mass vials or as a solution in chloroform for handling.
  • Well characterized: CAS 35418-58-7; formula C44H80NO8P.