Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-palmitoyl-2-glutaryl-sn-glycero-3-phosphocholine | 89947-79-5 | 99.9% | 609.73 g/mol | C29H56NO10P | 1 MG
SDP

Catalog No. 5000209089
Encompass_Preferred

1-palmitoyl-2-glutaryl-sn-glycero-3-phosphocholine (PGPC) is an oxidized phospholipid used in biochemical and cell-based research to study inflammation, cell signaling, and lipid-mediated biological responses. It is supplied as a purified, research-grade compound suitable for assay development and mechanistic studies.

  • High reported purity: 99.9%.
  • CAS number: 89947-79-5.
  • Molecular formula: C29H56NO10P; molecular weight: 609.73 g/mol.
  • Available in small quantities for laboratory research.
  • Suitable for biochemical and cell-based assays investigating lipid signaling and inflammation.

Catalog No. 50-002-09089 Supplier Medchemexpress LLC Supplier No. HYW0402551MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

1-palmitoyl-2-glutaryl-sn-glycero-3-phosphocholine (PGPC) is an oxidized phospholipid used in biochemical and cell-based research to study inflammation, cell signaling, and lipid-mediated biological responses. It is supplied as a purified, research-grade compound suitable for assay development and mechanistic studies.

  • High reported purity: 99.9%.
  • CAS number: 89947-79-5.
  • Molecular formula: C29H56NO10P; molecular weight: 609.73 g/mol.
  • Available in small quantities for laboratory research.
  • Suitable for biochemical and cell-based assays investigating lipid signaling and inflammation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.