Learn More
Medchemexpress LLC 1-palmitoyl-2-glutaryl-sn-glycero-3-phosphocholine | 89947-79-5 | MFCD05863933 | 99.9% | 609.73 g/mol | C29H56NO10P | 5 MG
SDP
Catalog No. 5000208927
Encompass_Preferred
1-palmitoyl-2-glutaryl-sn-glycero-3-phosphocholine (PGPC) is an oxidized phospholipid used as a biochemical research reagent to study lipid oxidation, inflammation, and membrane biology.
  • high purity suitable for analytical and biological assays.
  • Molecular formula C29H56NO10P and molecular weight 609.73 g/mol.
  • available in small package sizes for research workflows.
  • intended for research use only; not for human or clinical use.
Catalog No. 50-002-08927 Supplier Medchemexpress LLC Supplier No. HYW0402555MG
1-palmitoyl-2-glutaryl-sn-glycero-3-phosphocholine (PGPC) is an oxidized phospholipid used as a biochemical research reagent to study lipid oxidation, inflammation, and membrane biology.
  • high purity suitable for analytical and biological assays.
  • Molecular formula C29H56NO10P and molecular weight 609.73 g/mol.
  • available in small package sizes for research workflows.
  • intended for research use only; not for human or clinical use.