Learn More
Medchemexpress LLC 1-palmitoyl-2-hydroxy-sn-glycero-3-phospho-(1'-rac-glycerol) (sodium) | 326495-22-1 | 99.0% | 506.54 Da | C22H44NaO9P | 5 MG
SDP
Catalog No. 5000221765
Encompass_Preferred
1-Palmitoyl-2-hydroxy-sn-glycero-3-phospho-(1'-rac-glycerol) (sodium) is a palmitoyl lysophosphatidylglycerol (lyso-PG) lipid supplied as the sodium salt for biochemical and lipid research. It is a white to off-white solid with molecular formula C22H44NaO9P, molecular weight 506.54 Da, and nominal purity 99.0%. Recommended storage is -20°C (sealed, away from moisture); in solvent: -80°C up to 6 months or -20°C up to 1 month.
  • High purity (99.0%) suitable for analytical and preparative work
  • Lyso-PG lipid useful for membrane and signaling studies
  • White to off-white solid; easy to weigh and handle
  • Stable when stored sealed at -20°C; extended storage in solvent at -80°C
  • Available in small research quantities for laboratory experiments
Catalog No. 50-002-21765 Supplier Medchemexpress LLC Supplier No. HY1455075MG
1-Palmitoyl-2-hydroxy-sn-glycero-3-phospho-(1'-rac-glycerol) (sodium) is a palmitoyl lysophosphatidylglycerol (lyso-PG) lipid supplied as the sodium salt for biochemical and lipid research. It is a white to off-white solid with molecular formula C22H44NaO9P, molecular weight 506.54 Da, and nominal purity 99.0%. Recommended storage is -20°C (sealed, away from moisture); in solvent: -80°C up to 6 months or -20°C up to 1 month.
  • High purity (99.0%) suitable for analytical and preparative work
  • Lyso-PG lipid useful for membrane and signaling studies
  • White to off-white solid; easy to weigh and handle
  • Stable when stored sealed at -20°C; extended storage in solvent at -80°C
  • Available in small research quantities for laboratory experiments