Learn More
Medchemexpress LLC 1-palmitoyl-2-hydroxy-sn-glycero-3-phospho-(1'-rac-glycerol) (sodium) | 326495-22-1 | MFCD00214310 | 99.0% | C22H44NaO9P | 10MG
SDP
Catalog No. 5000261867
Encompass_Preferred
1-Palmitoyl-2-hydroxy-sn-glycero-3-phospho-(1'-rac-glycerol) (sodium) is a lysophospholipid (16:0 Lyso PG) supplied as the sodium salt. It contains palmitic acid at the sn-1 position and is used to prepare micelles, liposomes, and model membranes for membrane biology and formulation studies. High purity and defined composition support reproducible experiments.
  • High purity (99.0%) for consistent experimental outcomes.
  • Sodium salt form suitable for aqueous dispersion and membrane model systems.
  • Compatible with micelle and liposome preparation for delivery and formulation research.
  • Well-characterized molecular formula and weight for accurate calculations.
  • Available in small research sizes for laboratory use.
Catalog No. 50-002-61867 Supplier Medchemexpress LLC Supplier No. HY14550710MG
1-Palmitoyl-2-hydroxy-sn-glycero-3-phospho-(1'-rac-glycerol) (sodium) is a lysophospholipid (16:0 Lyso PG) supplied as the sodium salt. It contains palmitic acid at the sn-1 position and is used to prepare micelles, liposomes, and model membranes for membrane biology and formulation studies. High purity and defined composition support reproducible experiments.
  • High purity (99.0%) for consistent experimental outcomes.
  • Sodium salt form suitable for aqueous dispersion and membrane model systems.
  • Compatible with micelle and liposome preparation for delivery and formulation research.
  • Well-characterized molecular formula and weight for accurate calculations.
  • Available in small research sizes for laboratory use.