Learn More
Medchemexpress LLC 1-Palmitoyl-2-linoleoyl-sn-glycero-3-PC | 17708-90-6 | 99.0% | 758.06 | 5 MG
SDP
Catalog No. 5000391661
Encompass_Preferred
1-Palmitoyl-2-linoleoyl-sn-glycero-3-PC (PLPC) is a phospholipid used as a major component in lipid bilayer system models. It can serve as an untreated control, mixed with oxidized lipids such as 1-palmitoyl-2-(13-hydroxy-9,11-octadecanedienoyl)-lecithin (OHPLPC) and 1-palmitoyl-2-(13-hydroperoxy-9,11-octadecanedienoyl)-lecithin (HpPLPC), to study the effects of oxidation on membrane properties.
  • Used as a major component in lipid bilayer system models.
  • Can serve as an untreated control to study the effects of oxidation on membrane properties when mixed with oxidized lipids.
Catalog No. 50-003-91661 Supplier Medchemexpress LLC Supplier No. HY1650295MG
1-Palmitoyl-2-linoleoyl-sn-glycero-3-PC (PLPC) is a phospholipid used as a major component in lipid bilayer system models. It can serve as an untreated control, mixed with oxidized lipids such as 1-palmitoyl-2-(13-hydroxy-9,11-octadecanedienoyl)-lecithin (OHPLPC) and 1-palmitoyl-2-(13-hydroperoxy-9,11-octadecanedienoyl)-lecithin (HpPLPC), to study the effects of oxidation on membrane properties.
  • Used as a major component in lipid bilayer system models.
  • Can serve as an untreated control to study the effects of oxidation on membrane properties when mixed with oxidized lipids.