Learn More
Medchemexpress LLC 1-palmitoyl-2-linoleoyl-sn-glycero-3-phosphocholine | 17708-90-6 | 99.0% | 758.06 g/mol | C42H80NO8P | 10 MG
SDP
Catalog No. 5000289564
Encompass_Preferred
1-Palmitoyl-2-linoleoyl-sn-glycero-3-phosphocholine (PLPC) is a phosphatidylcholine lipid commonly used in membrane, liposome, and biochemical assay research. Supplied as a chloroform solution at research-grade purity, it is intended for formulation studies and analytical applications.
  • Phosphatidylcholine lipid for membrane and liposome studies.
  • High purity, 99.0%, suitable for analytical and preparative work.
  • Provided as a solution in chloroform for immediate use in formulations.
  • Recommended storage at -20°C, stable for up to 2 years in solution.
  • Available in small research quantities, including 5 mg and 10 mg options.
Catalog No. 50-002-89564 Supplier Medchemexpress LLC Supplier No. HY16502910MG
1-Palmitoyl-2-linoleoyl-sn-glycero-3-phosphocholine (PLPC) is a phosphatidylcholine lipid commonly used in membrane, liposome, and biochemical assay research. Supplied as a chloroform solution at research-grade purity, it is intended for formulation studies and analytical applications.
  • Phosphatidylcholine lipid for membrane and liposome studies.
  • High purity, 99.0%, suitable for analytical and preparative work.
  • Provided as a solution in chloroform for immediate use in formulations.
  • Recommended storage at -20°C, stable for up to 2 years in solution.
  • Available in small research quantities, including 5 mg and 10 mg options.