Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-palmitoyl-2-linoleoyl-sn-glycero-3-phosphoethanolamine | 26662-95-3 | 98.0% | 1 MG
SDP

Catalog No. 5000233991
Encompass_Preferred

1-Palmitoyl-2-linoleoyl-sn-glycero-3-PE is a phosphatidylethanolamine lipid reagent used in research to facilitate cellular uptake of nucleic acids such as small RNA. It is supplied as a powder and should be stored cold to maintain stability.

  • Phosphatidylethanolamine lipid that can assist nucleic acid delivery into cells
  • Cas 26662-95-3
  • Molecular formula C39H74NO8P, molecular weight 715.98
  • Purity 98.0%
  • Available in small research pack sizes, including 1 mg
  • Store powder at -20°C; in solvent store at -80°C for long-term

Catalog No. 50-002-33991 Supplier Medchemexpress LLC Supplier No. HY1209911MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Palmitoyl-2-linoleoyl-sn-glycero-3-PE is a phosphatidylethanolamine lipid reagent used in research to facilitate cellular uptake of nucleic acids such as small RNA. It is supplied as a powder and should be stored cold to maintain stability.

  • Phosphatidylethanolamine lipid that can assist nucleic acid delivery into cells
  • Cas 26662-95-3
  • Molecular formula C39H74NO8P, molecular weight 715.98
  • Purity 98.0%
  • Available in small research pack sizes, including 1 mg
  • Store powder at -20°C; in solvent store at -80°C for long-term

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.