Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-palmitoyl-2-linoleoyl-sn-glycero-3-phosphoethanolamine | 26662-95-3 | 98.0% | 10 MG
SDP

Catalog No. 5000231957
Encompass_Preferred

1-palmitoyl-2-linoleoyl-sn-glycero-3-phosphoethanolamine is a phosphatidylethanolamine lipid used in research to facilitate cellular uptake of nucleic acids and to support formulation of lipid-based delivery systems. The material is supplied as a white to off-white solid with high purity for biochemical and lipid formulation studies.

  • Purity 98.0%.
  • Molecular weight 715.98 g/mol.
  • Chemical formula C39H74NO8P.
  • Appearance white to off-white solid.
  • Intended for research use only.
  • Available in small sizes suitable for formulation studies (1 mg-100 mg).

Catalog No. 50-002-31957 Supplier Medchemexpress LLC Supplier No. HY12099110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-palmitoyl-2-linoleoyl-sn-glycero-3-phosphoethanolamine is a phosphatidylethanolamine lipid used in research to facilitate cellular uptake of nucleic acids and to support formulation of lipid-based delivery systems. The material is supplied as a white to off-white solid with high purity for biochemical and lipid formulation studies.

  • Purity 98.0%.
  • Molecular weight 715.98 g/mol.
  • Chemical formula C39H74NO8P.
  • Appearance white to off-white solid.
  • Intended for research use only.
  • Available in small sizes suitable for formulation studies (1 mg-100 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.