Learn More
Medchemexpress LLC 1-palmitoyl-2-linoleoyl-sn-glycero-3-phosphoethanolamine | 26662-95-3 | 98.0% | 25 MG
SDP
Catalog No. 5000231958
Encompass_Preferred
1-Palmitoyl-2-linoleoyl-sn-glycero-3-PE is a glycerophospholipid (phosphatidylethanolamine) used in liposome formulation and nucleic-acid delivery research. It is supplied as a powder and is documented to be stable at -20°C when stored dry, with defined stability periods in solvent.
  • Promotes nucleic acid adsorption and cellular delivery
  • Used in liposome and membrane formulation studies
  • Provided as a powder with cold-storage stability
  • Molecular weight 715.98 g/mol and formula C39H74NO8P
  • Intended for research use only
Catalog No. 50-002-31958 Supplier Medchemexpress LLC Supplier No. HY12099125MG
1-Palmitoyl-2-linoleoyl-sn-glycero-3-PE is a glycerophospholipid (phosphatidylethanolamine) used in liposome formulation and nucleic-acid delivery research. It is supplied as a powder and is documented to be stable at -20°C when stored dry, with defined stability periods in solvent.
  • Promotes nucleic acid adsorption and cellular delivery
  • Used in liposome and membrane formulation studies
  • Provided as a powder with cold-storage stability
  • Molecular weight 715.98 g/mol and formula C39H74NO8P
  • Intended for research use only