Learn More
Medchemexpress LLC 1-Palmitoyl-2-oleoyl-3-bromopropanediol | 80749-30-0 | 5 MG
SDP
Catalog No. 5000391273
Encompass_Preferred
1-Palmitoyl-2-oleoyl-3-bromopropanediol is a solid chemical identified for laboratory use and the manufacture of substances. It is stable under recommended storage conditions and is intended for research use only by experienced personnel in appropriately equipped facilities.
  • Intended for laboratory use
  • Stable under recommended storage conditions
  • Should be handled by suitably qualified scientists
  • Store in a cool, well-ventilated area
  • Keep away from direct sunlight and ignition sources
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-003-91273 Supplier Medchemexpress LLC Supplier No. HY1657325MG
1-Palmitoyl-2-oleoyl-3-bromopropanediol is a solid chemical identified for laboratory use and the manufacture of substances. It is stable under recommended storage conditions and is intended for research use only by experienced personnel in appropriately equipped facilities.
  • Intended for laboratory use
  • Stable under recommended storage conditions
  • Should be handled by suitably qualified scientists
  • Store in a cool, well-ventilated area
  • Keep away from direct sunlight and ignition sources
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month