Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Palmitoyl-2-oleoyl-3-bromopropanediol | 80749-30-0 | 99.0% | 657.85 g·mol⁻¹ | C37H69BrO4 | 10 MG
SDP

Catalog No. 5000289003
Encompass_Preferred

1-Palmitoyl-2-oleoyl-3-bromopropanediol is a brominated diacylglycerol ester supplied for research use. It is provided as a purified compound for biochemical studies, lipid modification research, and chemical synthesis applications. The compound is for research use only and not for human or clinical use.

  • Brominated diacylglycerol structure suitable for lipid-related studies
  • High purity: 99.0%
  • Molecular weight 657.85 g·mol⁻¹; chemical formula C37H69BrO4
  • Available in small-scale quantities (for example, 1 mg, 5 mg, 10 mg)
  • Store at 4°C and protect from light; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month)

Catalog No. 50-002-89003 Supplier Medchemexpress LLC Supplier No. HY16573210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Palmitoyl-2-oleoyl-3-bromopropanediol is a brominated diacylglycerol ester supplied for research use. It is provided as a purified compound for biochemical studies, lipid modification research, and chemical synthesis applications. The compound is for research use only and not for human or clinical use.

  • Brominated diacylglycerol structure suitable for lipid-related studies
  • High purity: 99.0%
  • Molecular weight 657.85 g·mol⁻¹; chemical formula C37H69BrO4
  • Available in small-scale quantities (for example, 1 mg, 5 mg, 10 mg)
  • Store at 4°C and protect from light; in solvent store at -80°C (up to 6 months) or -20°C (up to 1 month)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.