Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-palmitoyl-2-oleoyl-3-linoleoyl-rac-glycerol | 2680-59-3 | MFCD00214386 | >=98.0% | 857.38 | C55H100O6 | 5 MG
SDP

Catalog No. 5000285267
Encompass_Preferred

1-Palmitoyl-2-oleoyl-3-linoleoyl-rac-glycerol is a triacylglycerol (triglyceride) used as a biochemical reagent in lipid research and analytical applications. It serves as a representative mixed fatty acid glyceride and can be used as a reference reagent or standard in triglyceride analysis. For research use only; consult the certificate of analysis for lot-specific purity and storage instructions.

  • Triacylglycerol reagent for lipid research and assays.
  • Useful as a reference standard for triglyceride analysis.
  • Contains palmitic (16:0), oleic (18:1), and linoleic (18:2) fatty acid residues.
  • Molecular weight 857.38 g/mol; CAS 2680-59-3.
  • Certificate of analysis provides purity (≥98%) and storage guidance.

Catalog No. 50-002-85267 Supplier Medchemexpress LLC Supplier No. HYW5883155MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Palmitoyl-2-oleoyl-3-linoleoyl-rac-glycerol is a triacylglycerol (triglyceride) used as a biochemical reagent in lipid research and analytical applications. It serves as a representative mixed fatty acid glyceride and can be used as a reference reagent or standard in triglyceride analysis. For research use only; consult the certificate of analysis for lot-specific purity and storage instructions.

  • Triacylglycerol reagent for lipid research and assays.
  • Useful as a reference standard for triglyceride analysis.
  • Contains palmitic (16:0), oleic (18:1), and linoleic (18:2) fatty acid residues.
  • Molecular weight 857.38 g/mol; CAS 2680-59-3.
  • Certificate of analysis provides purity (≥98%) and storage guidance.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.