Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-palmitoyl-2-oleoyl-3-linoleoyl-rac-glycerol | 2680-59-3 | MFCD00214386 | >=98.0% | 857.38 g·mol⁻¹ | C55H100O6 | 10 MG
SDP

Catalog No. 5000284532
Encompass_Preferred

1-Palmitoyl-2-oleoyl-3-linoleoyl-rac-glycerol is a triacylglycerol reagent supplied for biochemical and lipid research, and for use as an analytical standard in assay development and lipid studies.

  • Triacylglycerol reagent used for biochemical assays and lipid research.
  • Molecular formula: C55H100O6.
  • Molecular weight: 857.38 g·mol⁻¹.
  • CAS number: 2680-59-3.
  • Purity: ≥98.0% as listed in compound records.
  • Provided as a small-mass research quantity for analytical use.

Catalog No. 50-002-84532 Supplier Medchemexpress LLC Supplier No. HYW58831510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Palmitoyl-2-oleoyl-3-linoleoyl-rac-glycerol is a triacylglycerol reagent supplied for biochemical and lipid research, and for use as an analytical standard in assay development and lipid studies.

  • Triacylglycerol reagent used for biochemical assays and lipid research.
  • Molecular formula: C55H100O6.
  • Molecular weight: 857.38 g·mol⁻¹.
  • CAS number: 2680-59-3.
  • Purity: ≥98.0% as listed in compound records.
  • Provided as a small-mass research quantity for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.