Learn More
Avanti Polar Lipids 1-PALMITOYL-2-OLEOYL-SN, 200MG
Catalog No. NC9218208
Encompass
1-Palmitoyl-2-Oleoyl-sn-Glycero-3-Phosphocholine, Hygroscopic as lyophilized powder, C 42 H&subscript_begin82 NO8P, Percent composition: C 66.37% H 10.87% N 1.84% O 16.84% P 4.08%, 200mg
Catalog No. NC9218208 Supplier Avanti Polar Lipids Supplier No. 850457P200MG
1-Palmitoyl-2-Oleoyl-sn-Glycero-3-Phosphocholine, Hygroscopic as lyophilized powder, C 42 H&subscript_begin82 NO8P, Percent composition: C 66.37% H 10.87% N 1.84% O 16.84% P 4.08%, 200mg