Learn More
Medchemexpress LLC HY-130462 5mg Medchemexpress, 1-Palmitoyl-2-oleoyl-sn-glycero-3-PC CAS:26853-31-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1304625MG
Encompass_Preferred
Medchemexpress, HY-130462 5mg 1-Palmitoyl-2-oleoyl-sn-glycero-3-PC CAS:26853-31-6 1-Palmitoyl-2-oleoyl-sn-glycero-3-PC (POPC), a phospholipid, is a major component of biological membranes. 1-Palmitoyl-2-oleoyl-sn-glycero-3-PC is used for the preparation of liposomes and studying the properties of lipid bilayers. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0208