Learn More
TARGETMOL CHEMICALS INC 1-Palmitoyl-2-oleoyl-sn-glycer
SDP
Supplier:  TARGETMOL CHEMICALS INC T19085100MG
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. 1-Palmitoyl-2-oleoyl-sn-glycero-3-PC (Palmitoyloleoylphosphatidylcholine) is a phospholipid commonly found in cell membranes, with cholesterol compositions ranging from 0 to 60 mol%. Purity 99.71%
Catalog No. 50-287-8358