Learn More
Medchemexpress LLC 1-Palmitoyl-2-oleoyl-sn-glycero-3-PC-d31 | 179093-76-6 | ≥99.0% | 1 MG
SDP
Catalog No. 5000339791
Encompass_Preferred
1-Palmitoyl-2-oleoyl-sn-glycero-3-PC-d31 is a deuterium-labeled version of 1-Palmitoyl-2-oleoyl-sn-glycero-3-PC (POPC). POPC is a phospholipid and a major component of biological membranes. The deuterium labeling allows this compound to be used as a tracer and as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs. The product is for research use only.
  • Deuterium labeled
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-39791 Supplier Medchemexpress LLC Supplier No. HY130462S1MG
1-Palmitoyl-2-oleoyl-sn-glycero-3-PC-d31 is a deuterium-labeled version of 1-Palmitoyl-2-oleoyl-sn-glycero-3-PC (POPC). POPC is a phospholipid and a major component of biological membranes. The deuterium labeling allows this compound to be used as a tracer and as an internal standard for quantitative analysis by techniques such as NMR, GC-MS, or LC-MS. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs. The product is for research use only.
  • Deuterium labeled
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS