Learn More
Medchemexpress LLC 1-Palmitoyl-2-oleoyl-sn-glycero-3-PC (POPC) | 26853-31-6 | 99.8% | 25 MG
SDP
Catalog No. 5000342295
Encompass_Preferred
1-Palmitoyl-2-oleoyl-sn-glycero-3-PC (POPC) is a phospholipid and a major component of biological membranes. It is used for the preparation of liposomes and studying the properties of lipid bilayers. POPC contains 16:0 and 18:1 fatty acids at the sn-1 and sn-2 positions, respectively.
  • Major component of biological membranes
  • Utilized in the preparation of liposomes
  • Used for studying the properties of lipid bilayers
  • Contains 16:0 and 18:1 fatty acids at the sn-1 and sn-2 positions
Catalog No. 50-003-42295 Supplier Medchemexpress LLC Supplier No. 25MGHY130462
1-Palmitoyl-2-oleoyl-sn-glycero-3-PC (POPC) is a phospholipid and a major component of biological membranes. It is used for the preparation of liposomes and studying the properties of lipid bilayers. POPC contains 16:0 and 18:1 fatty acids at the sn-1 and sn-2 positions, respectively.
  • Major component of biological membranes
  • Utilized in the preparation of liposomes
  • Used for studying the properties of lipid bilayers
  • Contains 16:0 and 18:1 fatty acids at the sn-1 and sn-2 positions