Learn More
Medchemexpress LLC 1-palmitoyl-2-oleoyl-sn-glycero-3-PE (1,2-POPE; 16:0-18:1 PE) | 26662-94-2 | MFCD00672608 | 99.5% | 718.00 g/mol | C39H76NO8P | 50 MG
SDP
Catalog No. 5000283883
Encompass_Preferred
1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphoethanolamine (1,2-POPE) is a phosphatidylethanolamine lipid commonly used in lipid nanoparticle (LNP) formulations to promote membrane fusion and enhance endosomal escape, improving cellular delivery efficiency of nucleic acid therapeutics such as mRNA. It is supplied as a white to off-white solid and is suitable for formulation and membrane biophysics research.
  • Promotes membrane fusion and facilitates endosomal escape.
  • Enhances lipid nanoparticle-mediated delivery of mRNA and other nucleic acids.
  • Induces hexagonal (HII) phase formation in acidic environments.
  • Available in multiple pack sizes to support formulation development.
  • High purity suitable for research applications.
Catalog No. 50-002-83883 Supplier Medchemexpress LLC Supplier No. HYW58386850MG
1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphoethanolamine (1,2-POPE) is a phosphatidylethanolamine lipid commonly used in lipid nanoparticle (LNP) formulations to promote membrane fusion and enhance endosomal escape, improving cellular delivery efficiency of nucleic acid therapeutics such as mRNA. It is supplied as a white to off-white solid and is suitable for formulation and membrane biophysics research.
  • Promotes membrane fusion and facilitates endosomal escape.
  • Enhances lipid nanoparticle-mediated delivery of mRNA and other nucleic acids.
  • Induces hexagonal (HII) phase formation in acidic environments.
  • Available in multiple pack sizes to support formulation development.
  • High purity suitable for research applications.