Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphate sodium | 169437-35-8 | MFCD00672609 | 99.0% | 696.91 g/mol | C37H70NaO8P | 5 MG
SDP

Catalog No. 5000222093
Encompass_Preferred

1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphate sodium is a high-purity phosphatidic acid (16:0-18:1) supplied as the monosodium salt for biochemical and membrane research. It is used in signaling, lipid metabolism, and liposome preparation studies where defined PA species are required.

  • Phosphatidic acid monosodium salt (16:0-18:1)
  • Molecular formula C37H70NaO8P
  • CAS 169437-35-8
  • Purity 99.0%
  • Solid reagent suitable for liposome and membrane assays
  • Common applications: signaling, membrane biology, lipid metabolism studies

Catalog No. 50-002-22093 Supplier Medchemexpress LLC Supplier No. HY134174A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphate sodium is a high-purity phosphatidic acid (16:0-18:1) supplied as the monosodium salt for biochemical and membrane research. It is used in signaling, lipid metabolism, and liposome preparation studies where defined PA species are required.

  • Phosphatidic acid monosodium salt (16:0-18:1)
  • Molecular formula C37H70NaO8P
  • CAS 169437-35-8
  • Purity 99.0%
  • Solid reagent suitable for liposome and membrane assays
  • Common applications: signaling, membrane biology, lipid metabolism studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.