Learn More
Medchemexpress LLC 1-palmitoyl-2-oleoyl-sn-glycero-3-phospho-L-serine sodium | 321863-21-2 | MFCD00674276 | 99.1% | 783.99 g·mol⁻¹ | C40H75NNaO10P | 25mg
SDP
Catalog No. 5000270148
Encompass_Preferred
2-Oleoyl-1-palmitoyl-sn-glycero-3-phospho-L-serine sodium is an anionic phospholipid with oleic acid (18 1) and palmitic acid (16 0) tails containing a carboxylic acid (COOH) and amine (NH2) in their head group It has been used in the preparation of lipid-mixing vesicles liposomes and artificial membranes
Catalog No. 50-002-70148 Supplier Medchemexpress LLC Supplier No. HYW71452425MG
2-Oleoyl-1-palmitoyl-sn-glycero-3-phospho-L-serine sodium is an anionic phospholipid with oleic acid (18 1) and palmitic acid (16 0) tails containing a carboxylic acid (COOH) and amine (NH2) in their head group It has been used in the preparation of lipid-mixing vesicles liposomes and artificial membranes