Learn More
Medchemexpress LLC 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphoethanolamine | 26662-94-2 | MFCD00672608 | 99.5% | 718.00 | C39H76NO8P | 25 MG
SDP
Catalog No. 5000283882
Encompass_Preferred
1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphoethanolamine is a phosphatidylethanolamine lipid used in membrane biophysics and lipid nanoparticle research. It promotes hexagonal phase formation and enhances membrane fusion, which can improve endosomal escape and cellular delivery of nucleic acid therapeutics.
  • Promotes hexagonal (HII) lipid phase formation under acidic conditions.
  • Facilitates membrane fusion and enhances endosomal escape.
  • Useful for lipid nanoparticle formulation and mRNA and gene delivery research.
  • High purity suitable for biochemical and biophysical assays.
  • Stable with defined storage recommendations for powder and in solvent.
Catalog No. 50-002-83882 Supplier Medchemexpress LLC Supplier No. HYW58386825MG
1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphoethanolamine is a phosphatidylethanolamine lipid used in membrane biophysics and lipid nanoparticle research. It promotes hexagonal phase formation and enhances membrane fusion, which can improve endosomal escape and cellular delivery of nucleic acid therapeutics.
  • Promotes hexagonal (HII) lipid phase formation under acidic conditions.
  • Facilitates membrane fusion and enhances endosomal escape.
  • Useful for lipid nanoparticle formulation and mRNA and gene delivery research.
  • High purity suitable for biochemical and biophysical assays.
  • Stable with defined storage recommendations for powder and in solvent.