Learn More
Medchemexpress LLC 1-palmitoyl-2-oleoyl-sn-glycerol, 1-oleoyl-3-palmitoylglycerol (1:1) | 00-00-0 | 97.2% | 1189.90 g·mol⁻¹ | C74H140O10 | 5 MG
SDP
Catalog No. 5000222043
Encompass_Preferred
1-Palmitoyl-2-oleoyl-sn-glycerol, 1-oleoyl-3-palmitoylglycerol (1:1) is a 1:1 mixture of two diacylglycerol isomers. It is a biologically active diacylglycerol (DAG) species and functions as a hypoxia-inducible factor (HIF) inducer, provided as a research reagent for studies of DAG signaling and HIF-related pathways.
  • Major active component of diacylglycerol (DAG).
  • Acts as a hypoxia-inducible factor (HIF) inducer.
  • Supplied as a purified solid suitable for research use.
  • High reported purity (97.2%).
  • Available in small pre-weighed pack sizes for convenient handling.
Catalog No. 50-002-22043 Supplier Medchemexpress LLC Supplier No. HY134200A5MG
1-Palmitoyl-2-oleoyl-sn-glycerol, 1-oleoyl-3-palmitoylglycerol (1:1) is a 1:1 mixture of two diacylglycerol isomers. It is a biologically active diacylglycerol (DAG) species and functions as a hypoxia-inducible factor (HIF) inducer, provided as a research reagent for studies of DAG signaling and HIF-related pathways.
  • Major active component of diacylglycerol (DAG).
  • Acts as a hypoxia-inducible factor (HIF) inducer.
  • Supplied as a purified solid suitable for research use.
  • High reported purity (97.2%).
  • Available in small pre-weighed pack sizes for convenient handling.