Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-palmitoyl-2-oleoyl-sn-glycerol | 29541-66-0 | 99.6% | 594.95 g/mol | C37H70O5 | 5 MG
SDP

Catalog No. 5000223458
Encompass_Preferred

1-Palmitoyl-2-oleoyl-sn-glycerol is an endogenous diacylglycerol metabolite and a major component of the hormogonium-inducing factor (HIF)-1. Supplied for research use only, it is commonly used in studies of lipid signaling, membrane interactions, and biochemical assays.

  • CAS number 29541-66-0.
  • Molecular formula C37H70O5; molecular weight 594.95 g/mol.
  • High purity (~99.6%) as reported by manufacturer.
  • Available as 5 mg neat and as DMSO solutions for immediate use.
  • Storage: pure form -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Certificate of analysis, datasheet, and SDS available.

Catalog No. 50-002-23458 Supplier Medchemexpress LLC Supplier No. HY1342005MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

1-Palmitoyl-2-oleoyl-sn-glycerol is an endogenous diacylglycerol metabolite and a major component of the hormogonium-inducing factor (HIF)-1. Supplied for research use only, it is commonly used in studies of lipid signaling, membrane interactions, and biochemical assays.

  • CAS number 29541-66-0.
  • Molecular formula C37H70O5; molecular weight 594.95 g/mol.
  • High purity (~99.6%) as reported by manufacturer.
  • Available as 5 mg neat and as DMSO solutions for immediate use.
  • Storage: pure form -20°C; in solvent -80°C (6 months) or -20°C (1 month).
  • Certificate of analysis, datasheet, and SDS available.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.