Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-palmitoyl-2-oleoyl-sn-glycerol | 29541-66-0 | MFCD00674221 | 99.9% | C37H70O5 | 10MG
SDP

Catalog No. 5000259467
Encompass_Preferred

1-Palmitoyl-2-oleoyl-sn-glycerol is an endogenous 1,2-diacyl-sn-glycerol supplied as a research-grade lipid standard for studies of diacylglycerol-mediated signaling, membrane biology, and analytical reference. Provided in high purity, it is suitable for preparation of stock solutions and use as a reference in biochemical and analytical workflows.

  • Endogenous diacylglycerol lipid involved in cellular signaling.
  • High purity (99.9%).
  • Suitable as a biochemical standard for lipid signaling assays and analytical reference.
  • Available as small quantities and as a DMSO solution for convenient stock preparation.
  • Chemical formula C37H70O5 and molecular weight 594.95.
  • Storage: pure form -20°C; in solvent -80°C for 6 months or -20°C for 1 month.

Catalog No. 50-002-59467 Supplier Medchemexpress LLC Supplier No. HY13420010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Palmitoyl-2-oleoyl-sn-glycerol is an endogenous 1,2-diacyl-sn-glycerol supplied as a research-grade lipid standard for studies of diacylglycerol-mediated signaling, membrane biology, and analytical reference. Provided in high purity, it is suitable for preparation of stock solutions and use as a reference in biochemical and analytical workflows.

  • Endogenous diacylglycerol lipid involved in cellular signaling.
  • High purity (99.9%).
  • Suitable as a biochemical standard for lipid signaling assays and analytical reference.
  • Available as small quantities and as a DMSO solution for convenient stock preparation.
  • Chemical formula C37H70O5 and molecular weight 594.95.
  • Storage: pure form -20°C; in solvent -80°C for 6 months or -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.