Learn More
Medchemexpress LLC 1-palmitoyl-2-stearoyl-sn-glycero-3-phosphocholine | 59403-51-9 | MFCD00674325 | 99.9% | 762.09 | C42H84NO8P | 50 MG
SDP
Catalog No. 5000281268
Encompass_Preferred
1-Palmitoyl-2-stearoyl-sn-glycero-3-phosphocholine is a synthetic phospholipid used in liposome formulation, membrane and lipid research, and related biochemical studies. Supplied as a powder for laboratory research use, it supports reproducible lipid preparations and analytical characterization.
  • High purity (99.9%).
  • Molecular formula C42H84NO8P; molecular weight 762.09.
  • Supplied as a powder suitable for liposome preparation.
  • Suggested storage: powder at -20°C to maintain stability.
  • Includes supporting documentation: data sheet, COA, and SDS.
Catalog No. 50-002-81268 Supplier Medchemexpress LLC Supplier No. HYW44095850MG
1-Palmitoyl-2-stearoyl-sn-glycero-3-phosphocholine is a synthetic phospholipid used in liposome formulation, membrane and lipid research, and related biochemical studies. Supplied as a powder for laboratory research use, it supports reproducible lipid preparations and analytical characterization.
  • High purity (99.9%).
  • Molecular formula C42H84NO8P; molecular weight 762.09.
  • Supplied as a powder suitable for liposome preparation.
  • Suggested storage: powder at -20°C to maintain stability.
  • Includes supporting documentation: data sheet, COA, and SDS.