Learn More
Medchemexpress LLC 1-palmitoyl-2-stearoyl-sn-glycero-3-phosphocholine | 59403-51-9 | MFCD00674325 | 99.9% | 762.09 g·mol⁻¹ | C42H84NO8P | 10 MG
SDP
Catalog No. 5000280499
Encompass_Preferred
1-Palmitoyl-2-stearoyl-sn-glycero-3-phosphocholine is an asymmetrical phosphatidylcholine used in liposome and membrane model research. It contains defined palmitoyl (C16:0) and stearoyl (C18:0) fatty acyl chains and the phosphate is esterified to choline. Supplied as a high-purity solid suitable for formulation, biophysical studies, and analytical applications.
  • Asymmetrical phospholipid with defined fatty acyl chains.
  • High purity suitable for analytical and formulation use.
  • Solid powder form for easy handling.
  • Recommended storage: powder -20°C for long-term stability.
  • Commonly used in liposome preparation and membrane model systems.
Catalog No. 50-002-80499 Supplier Medchemexpress LLC Supplier No. HYW44095810MG
Add to Cart
Add to Cart
1-Palmitoyl-2-stearoyl-sn-glycero-3-phosphocholine is an asymmetrical phosphatidylcholine used in liposome and membrane model research. It contains defined palmitoyl (C16:0) and stearoyl (C18:0) fatty acyl chains and the phosphate is esterified to choline. Supplied as a high-purity solid suitable for formulation, biophysical studies, and analytical applications.
  • Asymmetrical phospholipid with defined fatty acyl chains.
  • High purity suitable for analytical and formulation use.
  • Solid powder form for easy handling.
  • Recommended storage: powder -20°C for long-term stability.
  • Commonly used in liposome preparation and membrane model systems.