Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-palmitoyl-2-stearoyl-sn-glycero-3-phosphocholine | 59403-51-9 | MFCD00674325 | 99.9% | 762.09 g·mol⁻¹ | C42H84NO8P | 25 MG
SDP

Catalog No. 5000281175
Encompass_Preferred

1-Palmitoyl-2-stearoyl-sn-glycero-3-phosphocholine (PSPC) is a saturated phosphatidylcholine used as a reagent in lipid research and formulation. Supplied as a high-purity powder, it serves as a reference standard for analytical assays and as a building block for liposome and model membrane preparation when handled and stored under low-temperature conditions.

  • High purity: 99.9%.
  • Chemical formula C42H84NO8P; molecular weight 762.09 g·mol⁻¹.
  • Available in small research quantities, for example 25 MG.
  • Powder form for easy solubilization in suitable organic solvents.
  • Recommended storage: powder at -20°C; in solvent at -80°C for longer-term storage.
  • Applications include liposome formulation, membrane studies, and analytical standards.

Catalog No. 50-002-81175 Supplier Medchemexpress LLC Supplier No. HYW44095825MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Palmitoyl-2-stearoyl-sn-glycero-3-phosphocholine (PSPC) is a saturated phosphatidylcholine used as a reagent in lipid research and formulation. Supplied as a high-purity powder, it serves as a reference standard for analytical assays and as a building block for liposome and model membrane preparation when handled and stored under low-temperature conditions.

  • High purity: 99.9%.
  • Chemical formula C42H84NO8P; molecular weight 762.09 g·mol⁻¹.
  • Available in small research quantities, for example 25 MG.
  • Powder form for easy solubilization in suitable organic solvents.
  • Recommended storage: powder at -20°C; in solvent at -80°C for longer-term storage.
  • Applications include liposome formulation, membrane studies, and analytical standards.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.