Learn More
Medchemexpress LLC 1-palmitoyl-2-stearoyl-sn-glycero-3-phosphocholine | 59403-51-9 | MFCD00674325 | 99.9% | 762.09 g·mol⁻¹ | C42H84NO8P | 5 MG
SDP
Catalog No. 5000281650
Encompass_Preferred
1-Palmitoyl-2-stearoyl-sn-glycero-3-phosphocholine (PSPC) is an asymmetrical phosphatidylcholine lipid with palmitic acid at the sn-1 position and stearic acid at the sn-2 position. It is supplied as a high-purity research reagent for liposome formulation, model membranes, and membrane biophysics studies.
  • High purity (99.9%) suitable for research applications.
  • Molecular formula C42H84NO8P; molecular weight 762.09 g·mol⁻¹.
  • Used for liposome formulation and model membrane systems.
  • Available in small pack sizes for experimental work.
Catalog No. 50-002-81650 Supplier Medchemexpress LLC Supplier No. HYW4409585MG
1-Palmitoyl-2-stearoyl-sn-glycero-3-phosphocholine (PSPC) is an asymmetrical phosphatidylcholine lipid with palmitic acid at the sn-1 position and stearic acid at the sn-2 position. It is supplied as a high-purity research reagent for liposome formulation, model membranes, and membrane biophysics studies.
  • High purity (99.9%) suitable for research applications.
  • Molecular formula C42H84NO8P; molecular weight 762.09 g·mol⁻¹.
  • Used for liposome formulation and model membrane systems.
  • Available in small pack sizes for experimental work.