Learn More
Medchemexpress LLC 1-palmitoyl-sn-glycero-3-phosphocholine | 17364-16-8 | MFCD00036904 | 99.9% | 495.63 | C24H50NO7P | 25MG
SDP
Catalog No. 5000239070
Encompass_Preferred
1-Palmitoyl-sn-glycero-3-phosphocholine (16:0 Lyso PC) is a lysophosphatidylcholine lipid standard provided as a high-purity white solid for analytical and research applications. It is used as a reference material for method development, qualitative and quantitative analysis, and lipidomics studies, with documented stability and storage recommendations to ensure reproducibility.
  • High purity suitable for analytical methods.
  • Available in small-mass packs for trace-level experiments.
  • Compatible with LC, GC, and mass spectrometry workflows.
  • Stable when stored sealed at 4°C, protected from moisture and light.
  • Stock solutions remain stable at -80°C for up to six months.
Catalog No. 50-002-39070 Supplier Medchemexpress LLC Supplier No. HY12578325MG
1-Palmitoyl-sn-glycero-3-phosphocholine (16:0 Lyso PC) is a lysophosphatidylcholine lipid standard provided as a high-purity white solid for analytical and research applications. It is used as a reference material for method development, qualitative and quantitative analysis, and lipidomics studies, with documented stability and storage recommendations to ensure reproducibility.
  • High purity suitable for analytical methods.
  • Available in small-mass packs for trace-level experiments.
  • Compatible with LC, GC, and mass spectrometry workflows.
  • Stable when stored sealed at 4°C, protected from moisture and light.
  • Stock solutions remain stable at -80°C for up to six months.