Learn More
Medchemexpress LLC 1-palmitoyl-sn-glycero-3-phosphocholine | 17364-16-8 | MFCD00036904 | 99.9% | 495.63 g·mol⁻¹ | C24H50NO7P | 100MG
SDP
Catalog No. 5000235391
Encompass_Preferred
1-Palmitoyl-sn-glycero-3-phosphocholine is a lysophosphatidylcholine (16:0 Lyso PC) used in biochemical and cellular research. It is an abundant lysophospholipid with reported proinflammatory activity and is employed in studies of lipid signaling, inflammation, and atherosclerosis. The compound is supplied as a research-grade powder with molecular formula C24H50NO7P and molecular weight 495.63 g·mol⁻¹.
  • High purity (99.9%) suitable for analytical and in vitro studies.
  • Powder form convenient for storage and solution preparation.
  • Useful for studies of lipid signaling, inflammation, and atherosclerosis.
  • Soluble in common organic solvents for assay compatibility.
  • Available in multiple pack sizes to support different experimental scales.
Catalog No. 50-002-35391 Supplier Medchemexpress LLC Supplier No. HY125783100MG
1-Palmitoyl-sn-glycero-3-phosphocholine is a lysophosphatidylcholine (16:0 Lyso PC) used in biochemical and cellular research. It is an abundant lysophospholipid with reported proinflammatory activity and is employed in studies of lipid signaling, inflammation, and atherosclerosis. The compound is supplied as a research-grade powder with molecular formula C24H50NO7P and molecular weight 495.63 g·mol⁻¹.
  • High purity (99.9%) suitable for analytical and in vitro studies.
  • Powder form convenient for storage and solution preparation.
  • Useful for studies of lipid signaling, inflammation, and atherosclerosis.
  • Soluble in common organic solvents for assay compatibility.
  • Available in multiple pack sizes to support different experimental scales.