Learn More
Medchemexpress LLC 1-palmitoyl-sn-glycero-3-phosphocholine | 17364-16-8 | MFCD00036904 | 99.9% | 495.63 g/mol | C24H50NO7P | 50MG
SDP
Catalog No. 5000239072
Encompass_Preferred
1-Palmitoyl-sn-glycero-3-phosphocholine is a lysophosphatidylcholine used as a biochemical reagent and analytical standard in lipid research. It is an abundant lysophospholipid that can influence inflammatory responses and lipid signaling pathways, and is commonly employed in studies of membrane biology and atherosclerosis models.
  • High purity suitable for analytical and biochemical applications.
  • Commonly used as an analytical standard in lipidomics workflows.
  • Relevant for studies of inflammation, lipid signaling, and atherosclerosis.
  • Available in multiple small-scale quantities for laboratory experiments.
  • Stable when stored under recommended conditions for phospholipids.
Catalog No. 50-002-39072 Supplier Medchemexpress LLC Supplier No. HY12578350MG
1-Palmitoyl-sn-glycero-3-phosphocholine is a lysophosphatidylcholine used as a biochemical reagent and analytical standard in lipid research. It is an abundant lysophospholipid that can influence inflammatory responses and lipid signaling pathways, and is commonly employed in studies of membrane biology and atherosclerosis models.
  • High purity suitable for analytical and biochemical applications.
  • Commonly used as an analytical standard in lipidomics workflows.
  • Relevant for studies of inflammation, lipid signaling, and atherosclerosis.
  • Available in multiple small-scale quantities for laboratory experiments.
  • Stable when stored under recommended conditions for phospholipids.