Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Palmitoyl-sn-glycero-3-phosphocholine-d49 | 2315262-40-7 | >99.0% | 544.93 | C24HD49NO7P | 1MG
SDP

Catalog No. 5000236602
Encompass_Preferred

1-Palmitoyl-sn-glycero-3-phosphocholine-d49 is the deuterium-labeled isotopologue of 1-palmitoyl-sn-glycero-3-phosphocholine (16:0 Lyso PC). Supplied as a solid, it is used as an analytical reference and internal standard in lipidomics and mass spectrometry workflows to enable accurate quantitation of lysophosphatidylcholine species.

  • Isotopically labeled with 49 deuterium atoms (d49).
  • Chemical formula C24HD49NO7P.
  • Molecular weight 544.93.
  • CAS number 2315262-40-7.
  • Supplied as a solid in small pack sizes, suitable for analytical use.
  • Intended for use as an internal standard in LC-MS/MS and lipid analysis.

Catalog No. 50-002-36602 Supplier Medchemexpress LLC Supplier No. HY125783S21MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Palmitoyl-sn-glycero-3-phosphocholine-d49 is the deuterium-labeled isotopologue of 1-palmitoyl-sn-glycero-3-phosphocholine (16:0 Lyso PC). Supplied as a solid, it is used as an analytical reference and internal standard in lipidomics and mass spectrometry workflows to enable accurate quantitation of lysophosphatidylcholine species.

  • Isotopically labeled with 49 deuterium atoms (d49).
  • Chemical formula C24HD49NO7P.
  • Molecular weight 544.93.
  • CAS number 2315262-40-7.
  • Supplied as a solid in small pack sizes, suitable for analytical use.
  • Intended for use as an internal standard in LC-MS/MS and lipid analysis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.