Learn More
Medchemexpress LLC 1-palmitoyl-sn-glycero-3-phosphocholine-d9-1 | 182257-09-6 | >95.0% | 504.69 g/mol | C24H41D9NO7P | 1MG
SDP
Catalog No. 5000236456
Encompass_Preferred
1-Palmitoyl-sn-glycero-3-phosphocholine-d9-1 is a deuterium-labelled (d9) analogue of 1-palmitoyl lysophosphatidylcholine used as a stable isotope internal standard for lipidomics, quantitative mass spectrometry, and metabolic tracing. It supports accurate quantitation, retention time matching, and tracer studies, and is provided as a research reagent with listed purity ≥95% and molecular formula C24H41D9NO7P (MW 504.69 g/mol).
  • Provides a deuterated internal standard for accurate LC-MS/MS quantitation.
  • Matches chromatographic behavior of native lysophosphatidylcholine.
  • Enables tracer-based metabolic and flux studies.
  • High purity reduces matrix interference and improves reproducibility.
  • Convenient small-quantity packaging for standards preparation and method development.
Catalog No. 50-002-36456 Supplier Medchemexpress LLC Supplier No. HY125783S31MG
1-Palmitoyl-sn-glycero-3-phosphocholine-d9-1 is a deuterium-labelled (d9) analogue of 1-palmitoyl lysophosphatidylcholine used as a stable isotope internal standard for lipidomics, quantitative mass spectrometry, and metabolic tracing. It supports accurate quantitation, retention time matching, and tracer studies, and is provided as a research reagent with listed purity ≥95% and molecular formula C24H41D9NO7P (MW 504.69 g/mol).
  • Provides a deuterated internal standard for accurate LC-MS/MS quantitation.
  • Matches chromatographic behavior of native lysophosphatidylcholine.
  • Enables tracer-based metabolic and flux studies.
  • High purity reduces matrix interference and improves reproducibility.
  • Convenient small-quantity packaging for standards preparation and method development.