Learn More
Medchemexpress LLC 1-palmitoyl-sn-glycerol 3-phosphate sodium salt | 17618-08-5 | MFCD00674362 | 99.7% | 432.46 g/mol | C19H38NaO7P | 1 MG
SDP
Catalog No. 5000282079
Encompass_Preferred
1-Palmitoyl-sn-glycerol 3-phosphate sodium salt is a lysophosphatidic acid derivative used as a biochemical reagent for studies of glycerophospholipid metabolism and signaling. It is provided as a sodium salt solid intended for in vitro research and biochemical assays.
  • High purity suitable for sensitive analytical and biochemical experiments.
  • Supplied as a stable sodium salt for reliable handling and storage.
  • Useful for investigations of lipid metabolism and cell signaling pathways.
  • Available in research-scale quantities to support assay development.
  • Provided with product documentation such as datasheet and certificate of analysis.
Catalog No. 50-002-82079 Supplier Medchemexpress LLC Supplier No. HYW3373351MG
1-Palmitoyl-sn-glycerol 3-phosphate sodium salt is a lysophosphatidic acid derivative used as a biochemical reagent for studies of glycerophospholipid metabolism and signaling. It is provided as a sodium salt solid intended for in vitro research and biochemical assays.
  • High purity suitable for sensitive analytical and biochemical experiments.
  • Supplied as a stable sodium salt for reliable handling and storage.
  • Useful for investigations of lipid metabolism and cell signaling pathways.
  • Available in research-scale quantities to support assay development.
  • Provided with product documentation such as datasheet and certificate of analysis.