Learn More
Labtech Inc Dry Ice Condenser
Catalog No. NC1549720
Encompass_Preferred
High quality glass cold finger (dry ice) condenser. No chillers needed, utilizes dry ice for optimal efficiency.
Catalog No. NC1549720 Supplier Labtech Inc Supplier No. 50103I
High quality glass cold finger (dry ice) condenser. No chillers needed, utilizes dry ice for optimal efficiency.