Learn More
Medchemexpress LLC 1-Pentadecanol | 629-76-5 | 228.41 | 1 ML
SDP
Catalog No. 5000445420
Encompass_Preferred
1-Pentadecanol is a long-chain fatty alcohol with anti-comedone activity. It possesses moisturizing and hydrating properties, making it suitable for application in skin care products. Additionally, 1-Pentadecanol can be utilized in the production of surfactants, emulsifiers, and lubricants.
  • Anti-comedone activity
  • Moisturizing and hydrating properties
  • Suitable for skin care products
  • Can be used in production of surfactants
  • Can be used in production of emulsifiers
  • Can be used in production of lubricants
Catalog No. 50-004-45420 Supplier Medchemexpress LLC Supplier No. HYW00429510MM/1ML
1-Pentadecanol is a long-chain fatty alcohol with anti-comedone activity. It possesses moisturizing and hydrating properties, making it suitable for application in skin care products. Additionally, 1-Pentadecanol can be utilized in the production of surfactants, emulsifiers, and lubricants.
  • Anti-comedone activity
  • Moisturizing and hydrating properties
  • Suitable for skin care products
  • Can be used in production of surfactants
  • Can be used in production of emulsifiers
  • Can be used in production of lubricants