Learn More
Medchemexpress LLC 1-Pentadecanol | 629-76-5 | 228.41 | 100 G
SDP
Catalog No. 5000448794
Encompass_Preferred
1-Pentadecanol is a long-chain fatty alcohol known for its anti-comedone activity and its moisturizing and hydrating properties, making it suitable for skin care product formulations. It also finds applications in the production of surfactants, emulsifiers, and lubricants. This product is intended for research use only and is not for patient administration.
  • Long-chain fatty alcohol
  • Exhibits anti-comedone activity
  • Moisturizing and hydrating properties
  • Suitable for skin care product formulations
  • Utilized in surfactant production
  • Utilized in emulsifier production
  • Utilized in lubricant production
  • For research use only
Catalog No. 50-004-48794 Supplier Medchemexpress LLC Supplier No. HYW004295100G
1-Pentadecanol is a long-chain fatty alcohol known for its anti-comedone activity and its moisturizing and hydrating properties, making it suitable for skin care product formulations. It also finds applications in the production of surfactants, emulsifiers, and lubricants. This product is intended for research use only and is not for patient administration.
  • Long-chain fatty alcohol
  • Exhibits anti-comedone activity
  • Moisturizing and hydrating properties
  • Suitable for skin care product formulations
  • Utilized in surfactant production
  • Utilized in emulsifier production
  • Utilized in lubricant production
  • For research use only