Learn More
Medchemexpress LLC 1-Pentadecanol | 629-76-5 | 228.41 | 5 G
SDP
Catalog No. 5000445492
Encompass_Preferred
1-Pentadecanol is a long-chain fatty alcohol known for its anti-comedone activity and moisturizing and hydrating properties. It is suitable for skin care products and can be used in the production of surfactants, emulsifiers, and lubricants. It is also categorized as a biochemical assay reagent.
  • Exhibits anti-comedone activity
  • Possesses moisturizing and hydrating properties
  • Suitable for skin care products
  • Used in the production of surfactants, emulsifiers, and lubricants
  • Categorized as a biochemical assay reagent
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO (33.33 mg/mL with sonication)
  • Soluble in various in vivo dissolution protocols (≥ 2.5 mg/mL)
Catalog No. 50-004-45492 Supplier Medchemexpress LLC Supplier No. HYW0042955G
1-Pentadecanol is a long-chain fatty alcohol known for its anti-comedone activity and moisturizing and hydrating properties. It is suitable for skin care products and can be used in the production of surfactants, emulsifiers, and lubricants. It is also categorized as a biochemical assay reagent.
  • Exhibits anti-comedone activity
  • Possesses moisturizing and hydrating properties
  • Suitable for skin care products
  • Used in the production of surfactants, emulsifiers, and lubricants
  • Categorized as a biochemical assay reagent
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Soluble in DMSO (33.33 mg/mL with sonication)
  • Soluble in various in vivo dissolution protocols (≥ 2.5 mg/mL)