Learn More
Medchemexpress LLC 1-Pentadecanol | 629-76-5 | MFCD00004759 | 228.41 | 1 G
SDP
Catalog No. 5000445421
Encompass_Preferred
1-Pentadecanol is a long-chain fatty alcohol. It has anti-comedone activity, moisturizing and hydrating properties, and can be applied to skin care products. It can also be used in the production of surfactants, emulsifiers, and lubricants.
  • Has anti-comedone activity
  • Provides moisturizing and hydrating properties
  • Can be applied to skin care products
  • Used in the production of surfactants, emulsifiers, and lubricants
Catalog No. 50-004-45421 Supplier Medchemexpress LLC Supplier No. HYW0042951G
1-Pentadecanol is a long-chain fatty alcohol. It has anti-comedone activity, moisturizing and hydrating properties, and can be applied to skin care products. It can also be used in the production of surfactants, emulsifiers, and lubricants.
  • Has anti-comedone activity
  • Provides moisturizing and hydrating properties
  • Can be applied to skin care products
  • Used in the production of surfactants, emulsifiers, and lubricants