Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pentadecyne 95.0+%, TCI America™
SDP

Catalog No. P03565ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P03565ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P03565ML Supplier TCI America Supplier No. P03565ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 765-13-9
Molecular Formula C15H28
Molecular Weight (g/mol) 208.39
MDL Number MFCD00015081
InChI Key DONJGKADZJEXRJ-UHFFFAOYSA-N
Synonym 1-pentadecyne, 1-penatdecyne, pentadec-1-in, tridecylacetylene, 1-pentadecyne 1g, 1-pentadecyne,
PubChem CID 69825
IUPAC Name pentadec-1-yne
SMILES CCCCCCCCCCCCCC#C

Specifications

Color Yellow
Boiling Point 268°C
Quantity 5 mL
Formula Weight 208.39
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Pentadecyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.