Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Pentafluorophenyl)ethanol, TCI America™
SDP

Catalog No. P09255G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P09255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P09255G Supplier TCI America Supplier No. P09255G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 830-50-2
Molecular Formula C8H5F5O
Molecular Weight (g/mol) 212.12
MDL Number MFCD00004510
InChI Key WYUNHWKTLDBPLE-UHFFFAOYNA-N
Synonym 1-pentafluorophenyl ethanol, 1-2,3,4,5,6-pentafluorophenyl ethanol, 1-2,3,4,5,6-pentafluorophenyl ethan-1-ol, acmc-1bijr, 2,3,4,5,6-pentafluoro-alpha-methylbenzyl alcohol, 1-hydroxyethyl pentafluorobenzene, 1-pentafluorophenyl ethanol, 97, 2,3,4,5,6-pentafluoro-.alpha.-methylbenzyl alcohol, +/--alpha-methyl-2,3,4,5,6-pentafluorobenzyl alcohol, r-+--methyl-2,3,4,5,6-pentafluorobenzyl alcohol
PubChem CID 98288
IUPAC Name 1-(2,3,4,5,6-pentafluorophenyl)ethan-1-ol
SMILES CC(O)C1=C(F)C(F)=C(F)C(F)=C1F

Specifications

Quantity 5 g
Formula Weight 212.12
Physical Form Crystals
Chemical Name or Material 1-(Pentafluorophenyl)ethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.