Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pentanethiol, 97%

Catalog No. AAL0433014
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0433014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0433014 Supplier Thermo Scientific Chemicals Supplier No. L0433014
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 110-66-7 | C5H12S | 104.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-66-7
Molecular Formula C5H12S
Molecular Weight (g/mol) 104.21
MDL Number MFCD00004907
InChI Key ZRKMQKLGEQPLNS-UHFFFAOYSA-N
Synonym 1-pentanethiol, pentyl mercaptan, amyl mercaptan, n-amyl mercaptan, pentanethiol, amyl thioalcohol, pentalarm, amyl sulfhydrate, amyl hydrosulfide, pentylmercaptan
PubChem CID 8067
IUPAC Name pentane-1-thiol
SMILES CCCCCS

Specifications

Melting Point -76°C
Density 0.841
Boiling Point 125°C to 126°C
Flash Point 18°C (64°F)
Odor Stench
Refractive Index 1.446
Quantity 25 g
UN Number UN1111
Beilstein 1730979
Sensitivity Air Sensitive
Formula Weight 104.22
Percent Purity 97%
Chemical Name or Material 1-Pentanethiol
Show More Show Less
Hazard Category H225-H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H225-H302-H332-H315-H319-H335
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYL MERCAPTAN
EINECSNumber 203-789-1
RTECSNumber SA3150000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.