Learn More
Medchemexpress LLC 1-Pentanol, 2-amino-4-methyl-, (2S)- | 7533-40-6 | MFCD00063676 | 99.5% | 117.19 | C6H15NO | 25 MG
SDP
Catalog No. 5000303418
Encompass_Preferred
L-Leucinol ((+)-Leucinol) is a small-molecule research reagent that functions as a competitive aminopeptidase inhibitor (reported Ki ≈ 17 μM). It is provided as a high-purity compound intended for in vitro enzymatic studies and peptide synthesis.
  • High purity (≈99.5%) suitable for research applications.
  • Small molecule with molecular weight 117.19 and formula C6H15NO.
  • Competitive aminopeptidase inhibitor (Ki ≈ 17 μM) for enzymatic assays.
  • Available in multiple pack sizes including 25 mg.
  • Colorless to light yellow liquid with density 0.917 g/cm3.
  • Recommended storage: pure form -20°C (3 years) or 4°C (2 years).
Catalog No. 50-003-03418 Supplier Medchemexpress LLC Supplier No. 25MGHYW015876
L-Leucinol ((+)-Leucinol) is a small-molecule research reagent that functions as a competitive aminopeptidase inhibitor (reported Ki ≈ 17 μM). It is provided as a high-purity compound intended for in vitro enzymatic studies and peptide synthesis.
  • High purity (≈99.5%) suitable for research applications.
  • Small molecule with molecular weight 117.19 and formula C6H15NO.
  • Competitive aminopeptidase inhibitor (Ki ≈ 17 μM) for enzymatic assays.
  • Available in multiple pack sizes including 25 mg.
  • Colorless to light yellow liquid with density 0.917 g/cm3.
  • Recommended storage: pure form -20°C (3 years) or 4°C (2 years).