Learn More
Medchemexpress LLC 1-pentanol, 2-amino-4-methyl-, (2S)- | 7533-40-6 | MFCD00063676 | 99.5% | 117.19 g·mol⁻¹ | C6H15NO | 100 MG
SDP
Catalog No. 5000300980
Encompass_Preferred
L-Leucinol (CAS 7533-40-6) is an amino alcohol that acts as a competitive aminopeptidase inhibitor (reported Ki ≈ 17 μM). It is commonly used in in vitro enzymatic assays and serves as a building block or intermediate in peptide and small-molecule synthesis. A certificate of analysis for a referenced batch reports purity of 99.53% by GC.
  • Competitive aminopeptidase inhibitor with Ki ≈ 17 μM.
  • Suitable for in vitro enzymatic and biochemical assays.
  • Useful intermediate for peptide and small-molecule synthesis.
  • High purity typically reported (example: 99.53% by GC).
  • Available as solid and solution formulations for experimental flexibility.
Catalog No. 50-003-00980 Supplier Medchemexpress LLC Supplier No. HYW015876100MG
L-Leucinol (CAS 7533-40-6) is an amino alcohol that acts as a competitive aminopeptidase inhibitor (reported Ki ≈ 17 μM). It is commonly used in in vitro enzymatic assays and serves as a building block or intermediate in peptide and small-molecule synthesis. A certificate of analysis for a referenced batch reports purity of 99.53% by GC.
  • Competitive aminopeptidase inhibitor with Ki ≈ 17 μM.
  • Suitable for in vitro enzymatic and biochemical assays.
  • Useful intermediate for peptide and small-molecule synthesis.
  • High purity typically reported (example: 99.53% by GC).
  • Available as solid and solution formulations for experimental flexibility.