Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-pentanol, 2-amino-4-methyl-, (2S)- | 7533-40-6 | MFCD00063676 | 99.5% | 117.19 g/mol | C6H15NO | 500 MG
SDP

Catalog No. 5000303625
Encompass_Preferred

L-Leucinol ((+)-Leucinol) is the L-isomer of leucinol, a small-molecule amino alcohol used as a competitive aminopeptidase inhibitor and biochemical research reagent. Supplied as a high-purity liquid, it is suitable for enzymology, inhibitor characterization, and biochemical assay applications.

  • High purity (≈99.5%).
  • Acts as a competitive aminopeptidase inhibitor.
  • Molecular formula C6H15NO and molecular weight 117.19 g/mol.
  • Colorless to light yellow liquid; density 0.917 g/cm3.
  • Available in multiple pack sizes and solution formats for assay use.
  • Recommended storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-003-03625 Supplier Medchemexpress LLC Supplier No. HYW015876500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

L-Leucinol ((+)-Leucinol) is the L-isomer of leucinol, a small-molecule amino alcohol used as a competitive aminopeptidase inhibitor and biochemical research reagent. Supplied as a high-purity liquid, it is suitable for enzymology, inhibitor characterization, and biochemical assay applications.

  • High purity (≈99.5%).
  • Acts as a competitive aminopeptidase inhibitor.
  • Molecular formula C6H15NO and molecular weight 117.19 g/mol.
  • Colorless to light yellow liquid; density 0.917 g/cm3.
  • Available in multiple pack sizes and solution formats for assay use.
  • Recommended storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.