Learn More
Medchemexpress LLC 1-Pentanol, 4-methyl- | 626-89-1 | 102.18 | 1 G
SDP
Catalog No. 5000446421
Encompass_Preferred
4-Methyl-1-pentanol is a volatile aroma component found in red wine, often used in its production and blending. This colorless to light yellow liquid can act as an alcohol antagonist, counteracting the effects of ethanol and 1-butanol on cell-cell adhesion, and is utilized in studies related to fetal alcohol syndrome.
  • Volatile aroma component of red wine
  • Used in wine production and blending
  • Acts as an alcohol antagonist
  • Counteracts effects of ethanol and 1-butanol on cell-cell adhesion
  • Utilized in studies related to fetal alcohol syndrome
  • Density of 0.821 g/cm³
Catalog No. 50-004-46421 Supplier Medchemexpress LLC Supplier No. HYW0075111G
4-Methyl-1-pentanol is a volatile aroma component found in red wine, often used in its production and blending. This colorless to light yellow liquid can act as an alcohol antagonist, counteracting the effects of ethanol and 1-butanol on cell-cell adhesion, and is utilized in studies related to fetal alcohol syndrome.
  • Volatile aroma component of red wine
  • Used in wine production and blending
  • Acts as an alcohol antagonist
  • Counteracts effects of ethanol and 1-butanol on cell-cell adhesion
  • Utilized in studies related to fetal alcohol syndrome
  • Density of 0.821 g/cm³