Learn More
Medchemexpress LLC 1-pentanone, 1-(4-hydroxyphenyl)- | 2589-71-1 | MFCD00009719 | 100.0% | 178.23 g·mol⁻¹ | C11H14O2 | 25 G
SDP
Catalog No. 5000302794
Encompass_Preferred
4'-Hydroxyvalerophenone (4-pentanoylphenol) is a small-molecule research reagent that inhibits 17β-hydroxysteroid dehydrogenase (17β-HSD) with an IC50 of 60.52 μM. It is supplied as a white to off-white solid for biochemical assays and compound screening.
  • Inhibits 17β-hydroxysteroid dehydrogenase; IC50 = 60.52 μM.
  • High purity solid suitable for biochemical assays.
  • Molecular formula C11H14O2; molecular weight 178.23 g·mol⁻¹.
  • Available in multiple packaging sizes, including 25 g.
  • Stable as a powder under recommended storage conditions.
  • Supplied with a certificate of analysis for lot-specific quality data.
Catalog No. 50-003-02794 Supplier Medchemexpress LLC Supplier No. 25GHYW016604
Add to Cart
Add to Cart
4'-Hydroxyvalerophenone (4-pentanoylphenol) is a small-molecule research reagent that inhibits 17β-hydroxysteroid dehydrogenase (17β-HSD) with an IC50 of 60.52 μM. It is supplied as a white to off-white solid for biochemical assays and compound screening.
  • Inhibits 17β-hydroxysteroid dehydrogenase; IC50 = 60.52 μM.
  • High purity solid suitable for biochemical assays.
  • Molecular formula C11H14O2; molecular weight 178.23 g·mol⁻¹.
  • Available in multiple packaging sizes, including 25 g.
  • Stable as a powder under recommended storage conditions.
  • Supplied with a certificate of analysis for lot-specific quality data.